Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-BOC-4-methylaminopiperidine, 95%, Thermo Scientific™

Catalog No. AC454800050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454800050 5 g
AC454800010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC454800050 Supplier Thermo Scientific Chemicals Supplier No. 454800050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 147539-41-1
Molecular Formula C11H22N2O2
Molecular Weight (g/mol) 214.31
InChI Key CZYUGTLMFHDODF-UHFFFAOYSA-N
Synonym 1-boc-4-methylaminopiperidine, tert-butyl 4-methylamino piperidine-1-carboxylate, 1-boc-4-methylamino piperidine, 1-boc-4-methylaminopieridine, n-boc-4-methylaminopieridine, n-boc-4-methylaminopiperidine, 4-methylamino-piperidine-1-carboxylic acid tert-butyl ester, 4-methylamino piperidine, n1-boc protected, n-boc-4-n-methylamino piperidine, 4-n-methylamino-1-boc-piperidine
PubChem CID 15380702
IUPAC Name tert-butyl 4-(methylamino)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)NC

Specifications

Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Quantity 5 g
Formula Weight 214.31
Percent Purity 95%
Chemical Name or Material 1-BOC-4-methylaminopiperidine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.