accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1476-39-7

IUPAC Name: hydrogen pentane-1,5-diamine chloride
Synonyms: hydrogen pentane-1,5-diamine chloride
Molecular Weight (g/mol): 138.64
Molecular Formula: C5H15ClN2
InChi Key: RLNAIWYXIAJDTN-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCCCCCN
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)