Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Diaminopentane Dihydrochloride 98.0+%, TCI America™
SDP

Catalog No. D009925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D009925G 25 g
D00995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D009925G Supplier TCI America Supplier No. D009925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1476-39-7
Molecular Formula C5H16Cl2N2
Molecular Weight (g/mol) 175.097
MDL Number MFCD00012527
InChI Key FGNLEIGUMSBZQP-UHFFFAOYSA-N
Synonym Cadaverine Hydrochloride, Pentamethylenediamine Hydrochloride, 1,5-Pentanediamine Dihydrochloride
PubChem CID 80282
ChEBI CHEBI:64701
IUPAC Name pentane-1,5-diamine;dihydrochloride
SMILES C(CCN)CCN.Cl.Cl

Specifications

Melting Point 230°C
Color White-Yellow
Quantity 25 g
Formula Weight 175.10
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Diaminopentane Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.