accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaCH3OONH2OHOOOCHNOOHCH3CH3OOCH3OCH3CH3

CAS RN 1476-53-5

IUPAC Name: sodium 7-{[4-(carbamoyloxy)-3-hydroxy-5-methoxy-6,6-dimethyloxan-2-yl]oxy}-3-[4-hydroxy-3-(3-methylbut-2-en-1-yl)benzamido]-8-methyl-2,4-dioxo-3,4-dihydro-2H-1-benzopyran-3-ide
Synonyms: novobiocin
Molecular Weight (g/mol): 634.61
Molecular Formula: C31H35N2NaO11
InChi Key: AXOUUAINTJNFRS-UHFFFAOYNA-N
SMILES: [Na+].COC1C(OC(N)=O)C(O)C(OC2=CC=C3C(=O)[C-](NC(=O)C4=CC=C(O)C(CC=C(C)C)=C4)C(=O)OC3=C2C)OC1(C)C
Physical Form 
  • (2)
  • (7)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Type 
  • (5)
Color 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (5)
  • (2)
  • (7)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (5)
  • (3)
  • (2)