Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novobiocin sodium salt, 20 mg/ml in distilled water, sterile-filtered

Catalog No. AAJ67381AE
Change view
Click to view available options
Quantity:
50 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67381AE 100 mL
AAJ67381AD 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67381AE Supplier Thermo Scientific Chemicals Supplier No. J67381AE
Add to Cart

Novobiocin sodium salt, CAS # 1476-53-5, is a topoisomerase II inhibitor capable of inhibiting DNA synthesis. This compound is produced by Streptomyces nieveus and shows antibiotic activity. | CAS: 1476-53-5 | C31H35N2NaO11 | 634.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Novobiocin sodium salt, first isolated from Streptomyces nieveus, is a topoisomerase II inhibitor. It is a coumarin compound showing antibiotic activity
• Novobiocin sodium salt can inhibit DNA synthesis by inhibiting the topoisomerase II enzyme. Its antibacterial activity arises from competitive inhibition of the ATPase reaction catalyzed by the GyrB subunit

Application

• Novobiocin sodium salt is used to inhibit the activity of retroviral RNA-dependent DNA polymerase in laboratory experiments
• It is also used as an inhibitor of bacterial DNA gyrase and eukaryotic DNA topoisomerase.
• Novobiocin sodium salt shows anti-staphylococcal activity against methicillin-resistant Staphylococcus aureus, a gram-positive bacterium
• It permits the relaxation of negative DNA supercoils and has been used to generate positively supercoiled DNA
• This compound inhibits the chaperone activity of Hsp90

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1476-53-5
Molecular Formula C31H35N2NaO11
Molecular Weight (g/mol) 634.61
MDL Number MFCD00066541,MFCD00066541
InChI Key AXOUUAINTJNFRS-UHFFFAOYNA-N
PubChem CID 131673945
SMILES [Na+].COC1C(OC(N)=O)C(O)C(OC2=CC=C3C(=O)[C-](NC(=O)C4=CC=C(O)C(CC=C(C)C)=C4)C(=O)OC3=C2C)OC1(C)C

Specifications

Type Novobiocin Sodium Salt
Physical Form Liquid
Quantity 100 mL
IUPAC Name sodium 7-{[4-(carbamoyloxy)-3-hydroxy-5-methoxy-6,6-dimethyloxan-2-yl]oxy}-3-[4-hydroxy-3-(3-methylbut-2-en-1-yl)benzamido]-8-methyl-2,4-dioxo-3,4-dihydro-2 H-1-benzopyran-3-ide
Chemical Name or Material Novobiocin sodium salt, 20 mg/mL in distilled water, sterile-filtered
Hazard Category Warning
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 216-023-6
RTECSNumber RD5425000
TSCA No
storageNote1 -30°C to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.