accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NONHO

CAS RN 1476067-44-3

IUPAC Name: 4-{1-ethyl-5-oxa-8-azatricyclo[4.3.1.0³,⁸]decan-4-yl}quinolin-6-ol
Synonyms: 4-{1-ethyl-5-oxa-8-azatricyclo[4.3.1.0³,⁸]decan-4-yl}quinolin-6-ol
Molecular Weight (g/mol): 310.4
Molecular Formula: C19H22N2O2
InChi Key: DZHXBRDYIVRDPL-UHFFFAOYNA-N
SMILES: CCC12CC3C(OC(CN3C1)C2)C1=CC=NC2=CC=C(O)C=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results