Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,3S,5R,7R,8aS)-7-Ethylhexahydro-1-(6-hydroxy-4-quinolinyl)-3,7-methano-1H-pyrrolo[2,1-c][1,4]oxazine 98.0+%, TCI America™
SDP

Catalog No. E0974100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0974100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E0974100MG Supplier TCI America Supplier No. E0974100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1476067-44-3
Molecular Formula C19H22N2O2
Molecular Weight (g/mol) 310.397
InChI Key DZHXBRDYIVRDPL-DESPKHBRSA-N
Synonym alpha-Isocupreine, alpha-ICPN
PubChem CID 133554250
SMILES CCC12CC3CN(C1)C(C2)C(O3)C4=C5C=C(C=CC5=NC=C4)O

Specifications

Quantity 100 mg
Formula Weight 310.40
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (1R,3S,5R,7R,8aS)-7-Ethylhexahydro-1-(6-hydroxy-4-quinolinyl)-3,7-methano-1H-pyrrolo[2,1-c][1,4]oxaz
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.