CAS RN 147622-85-3
IUPAC Name:
4'-(3,4-difluorophenyl)-[1,1'-bi(cyclohexane)]-4-one
Synonyms:
4'-(3,4-difluorophenyl)-[1,1'-bi(cyclohexane)]-4-one
Molecular Weight (g/mol):
292.37
Molecular Formula:
C18H22F2O
InChi Key:
IUJMYVYPSSBPCX-UHFFFAOYSA-N
SMILES:
FC1=C(F)C=C(C=C1)C1CCC(CC1)C1CCC(=O)CC1
1
–
1
of
1
results
1
trans-4'-(3,4-Difluorophenyl)bicyclohexyl-4-one 98.0+%, TCI America™
CAS: 147622-85-3 Molecular Formula: C18H22F2O Molecular Weight (g/mol): 292.37 InChI Key: IUJMYVYPSSBPCX-UHFFFAOYSA-N PubChem CID: 11493045 IUPAC Name: 4-[4-(3,4-difluorophenyl)cyclohexyl]cyclohexan-1-one SMILES: C1CC(CCC1C2CCC(=O)CC2)C3=CC(=C(C=C3)F)F