Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4'-(3,4-Difluorophenyl)bicyclohexyl-4-one 98.0+%, TCI America™
SDP

Catalog No. D49115G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49115G 5 g
D49111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49115G Supplier TCI America Supplier No. D49115G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 147622-85-3
Molecular Formula C18H22F2O
Molecular Weight (g/mol) 292.37
InChI Key IUJMYVYPSSBPCX-UHFFFAOYSA-N
PubChem CID 11493045
IUPAC Name 4-[4-(3,4-difluorophenyl)cyclohexyl]cyclohexan-1-one
SMILES C1CC(CCC1C2CCC(=O)CC2)C3=CC(=C(C=C3)F)F

Specifications

Melting Point 77°C
Color White
Quantity 5 g
Formula Weight 292.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-4′-(3,4-Difluorophenyl)bicyclohexyl-4-one
EINECSNumber (4)-1815
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.