accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3O

CAS RN 14765-30-1

IUPAC Name: 2-(butan-2-yl)cyclohexan-1-one
Synonyms: 2-sec-butylcyclohexanone
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: RQXTZKGDMNIWJF-UHFFFAOYNA-N
SMILES: CCC(C)C1CCCCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results