Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-sec-Butylcyclohexanone (mixture of isomers) 98.0+%, TCI America™
SDP

Catalog No. B337125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B337125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B337125G Supplier TCI America Supplier No. B337125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14765-30-1
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00051454
InChI Key RQXTZKGDMNIWJF-UHFFFAOYSA-N
Synonym 2-sec-butylcyclohexanone, 2-sec-butyl cyclohexanone, cyclohexanone, 2-1-methylpropyl, 2-1-methylpropyl cyclohexanone, 2-sec-butylcyclohexan-1-one, cyclohexanone, 2-sec-butyl, butylcyclohexanone, o-sec, fema no. 3261, cyclohexanone, 2-sec-butyl-7ci,8ci, cyclohexanone,2-1-methylpropyl
PubChem CID 61771
IUPAC Name 2-butan-2-ylcyclohexan-1-one
SMILES CCC(C)C1CCCCC1=O

Specifications

Color Yellow
Boiling Point 78°C
Quantity 25 g
Formula Weight 154.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-sec-Butylcyclohexanone (mixture of isomers)
EINECSNumber (3)-2411
RTECSNumber GW1804000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.