accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNONONCH3CH3ClH3CNNONONCH3CH3Cl(E/Z)(E/Z)

CAS RN 147700-11-6

IUPAC Name: bis(8-[2-(3-chlorophenyl)ethenyl]-1,3,7-trimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione)
Synonyms: bis(8-(3-chlorostyryl)caffeine)
Molecular Weight (g/mol): 661.54
Molecular Formula: C32H30Cl2N8O4
InChi Key: YMUNFKYGBSLPLQ-UHFFFAOYSA-N
SMILES: CN1C(C=CC2=CC(Cl)=CC=C2)=NC2=C1C(=O)N(C)C(=O)N2C.CN1C(C=CC2=CC(Cl)=CC=C2)=NC2=C1C(=O)N(C)C(=O)N2C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results