CAS RN 147702-15-6
CAS RN 147702-15-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
Sigma Aldrich ™-(+)-2,2',6,6'-Tetramethoxy-4,4'-Bis(Di(3,5-Xylyl)Phosphino)-3,3'-Bipyridine
Sigma Aldrich ™-(+)-2,2',6,6'-Tetramethoxy-4,4'-Bis(Di(3,5-Xylyl)Phosphino)-3,3'-Bipyridine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.