accessibility menu, dialog, popup

UserName

Viewing profile as user:
POOHOOOOHHOONH(S)

CAS RN 147762-53-6

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-[4-(phosphonooxy)phenyl]propanoic acid
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-[4-(phosphonooxy)phenyl]propanoic acid
Molecular Weight (g/mol): 483.41
Molecular Formula: C24H22NO8P
InChi Key: AMXUJIHSMANWDW-QFIPXVFZSA-N
SMILES: OC(=O)[C@H](CC1=CC=C(OP(O)(O)=O)C=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)