accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 147762-89-8

IUPAC Name: (+)-1,1'-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]ferrocene
Synonyms: —
Molecular Weight (g/mol): 470.4
Molecular Formula: C26H40FeP2
InChi Key: —
SMILES: [Fe].CC[C@@H]1CC[C@@H](CC)P1c1cccc1.CC[C@@H]1CC[C@@H](CC)P1c1cccc1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results