Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-1,1'-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]ferrocene, 97+%, Thermo Scientific Chemicals

Catalog No. AAH6036403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6036403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6036403 Supplier Thermo Scientific Chemicals Supplier No. H6036403
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147762-89-8
Molecular Formula C26H40FeP2
Molecular Weight (g/mol) 470.40
MDL Number MFCD09265149
InChI Key MXOVWRYEUJWIFP-UHFFFAOYNA-N
Synonym +-1,1-bis 2r,5r-2,5-diethyl-1-phospholanyl ferrocene, 97+%
PubChem CID 131675863
SMILES [Fe].CC[C@@H]1CC[C@@H](CC)P1c1cccc1.CC[C@@H]1CC[C@@H](CC)P1c1cccc1

Specifications

Melting Point 85°C to 91°C
Quantity 1 g
Sensitivity Air Sensitive
Optical Rotation +380° (c=1 in Chloroform)
IUPAC Name carbanide;(2R,5R)-1-cyclopentyl-2,5-diethylphospholane;iron(2+)
Formula Weight 470.39
Percent Purity ≥97%
Chemical Name or Material (+)-1,1'-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]ferrocene
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.