accessibility menu, dialog, popup

UserName

Viewing profile as user:
FClN

CAS RN 1480-65-5

IUPAC Name: 5-chloro-2-fluoropyridine
Synonyms: 5-chloro-2-fluoropyridine
Molecular Weight (g/mol): 131.53
Molecular Formula: C5H3ClFN
InChi Key: ZULRQGBHWBQPFE-UHFFFAOYSA-N
SMILES: FC1=CC=C(Cl)C=N1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results