Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2-fluoropyridine 96.0+%, TCI America™
SDP

Catalog No. C23035G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23035G 5 g
C230325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23035G Supplier TCI America Supplier No. C23035G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1480-65-5
Molecular Formula C5H3ClFN
Molecular Weight (g/mol) 131.534
MDL Number MFCD04112513
InChI Key ZULRQGBHWBQPFE-UHFFFAOYSA-N
Synonym 2-fluoro-5-chloropyridine, 3-chloro-6-fluoropyridine, 5-chloro-2-fluoro-pyridine, 5-chloro-2-fluoro pyridine, pyridine, 5-chloro-2-fluoro, pubchem1186, acmc-1bwkp, 2-fluoro-5-chloro pyridine, ksc490s2r, 5-chloro-2-fluoropyridine
PubChem CID 345817
IUPAC Name 5-chloro-2-fluoropyridine
SMILES C1=CC(=NC=C1Cl)F

Specifications

Color Yellow
Boiling Point 149°C
Quantity 5 g
UN Number 1993
Formula Weight 131.53
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Chloro-2-fluoropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.