accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONH(S)NHNOONOOOOH

CAS RN 148083-64-1

IUPAC Name: (2S)-6-[(2,4-dinitrophenyl)amino]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Synonyms: (2S)-6-[(2,4-dinitrophenyl)amino]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanoic acid
Molecular Weight (g/mol): 534.53
Molecular Formula: C27H26N4O8
InChi Key: OENCMORJQAUAAJ-FQKVKQEKNA-N
SMILES: OC(=O)[C@H](CCCCNC1=C(C=C(C=C1)[N+]([O-])=O)[N+]([O-])=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)