accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHO

CAS RN 1481-93-2

IUPAC Name: 3,4-dihydro-2H-1-benzopyran-4-ol
Synonyms: 3,4-dihydro-2H-1-benzopyran-4-ol
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: MGSHXMOLUWTMGP-UHFFFAOYNA-N
SMILES: OC1CCOC2=CC=CC=C12
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)