accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1484-80-6

IUPAC Name: (2R)-2-ethylpiperidine
Synonyms: (2R)-2-ethylpiperidine
Molecular Weight (g/mol): 113.2
Molecular Formula: C7H15N
InChi Key: QBBKKFZGCDJDQK-SSDOTTSWSA-N
SMILES: CC[C@@H]1CCCCN1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)