Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylpiperidine 98.0+%, TCI America™
SDP

Catalog No. E030825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E030825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E030825ML Supplier TCI America Supplier No. E030825ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1484-80-6
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.204
MDL Number MFCD00005988
InChI Key QBBKKFZGCDJDQK-UHFFFAOYSA-N
Synonym piperidine, 2-ethyl, .alpha.-ethylpiperidine, 2-ethylpiperidin, 2-ethyl-piperidine, alpha-ethylpiperidine, 2-ethvlpiperidine, 2-ethyl-piperidin, 2-ethyl piperidine, pubchem6758, acmc-1bgny
PubChem CID 94205
IUPAC Name 2-ethylpiperidine
SMILES CCC1CCCCN1

Specifications

Color Yellow
Boiling Point 143°C
Quantity 25 mL
UN Number 1993
Formula Weight 113.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylpiperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.