accessibility menu, dialog, popup

UserName

Viewing profile as user:
KRuClClNClClClOK

CAS RN 14854-54-7

IUPAC Name: dipotassium pentachloro(nitroso)rutheniumbis(ylium)
Synonyms: dipotassium pentachloro(nitroso)rutheniumbis(ylium)
Molecular Weight (g/mol): 386.52
Molecular Formula: Cl5K2NORu
InChi Key: ZLWZLQOGXCFPCK-UHFFFAOYSA-I
SMILES: [K+].[K+].Cl[Ru++](Cl)(Cl)(Cl)(Cl)N=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results