accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14856-79-2

CH3SiH3CCH3HN

CAS RN 14856-79-2

IUPAC Name: benzyl(trimethylsilyl)amine
Synonyms: benzyl(trimethylsilyl)amine
Molecular Weight (g/mol): 179.34
Molecular Formula: C10H17NSi
InChi Key: OVONARZNQIKMGQ-UHFFFAOYSA-N
SMILES: C[Si](C)(C)NCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results