Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyltrimethylsilylamine 98.0+%, TCI America™
SDP

Catalog No. B17285G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B17285G Supplier TCI America Supplier No. B17285G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14856-79-2
Molecular Formula C10H17NSi
Molecular Weight (g/mol) 179.34
MDL Number MFCD00075505
InChI Key OVONARZNQIKMGQ-UHFFFAOYSA-N
Synonym (Benzylamino)trimethylsilane, N-(Trimethylsilyl)benzylamine
PubChem CID 519016
IUPAC Name benzyl(trimethylsilyl)amine
SMILES C[Si](C)(C)NCC1=CC=CC=C1

Specifications

Color Yellow
Quantity 5 g
UN Number 2920
Formula Weight 179.34
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material N-Benzyltrimethylsilylamine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.