CAS RN 1486-50-6
IUPAC Name:
4-(benzyloxy)benzoyl chloride
Synonyms:
4-(benzyloxy)benzoyl chloride
Molecular Weight (g/mol):
246.69
Molecular Formula:
C14H11ClO2
InChi Key:
ICEKEZSKMGHZNT-UHFFFAOYSA-N
SMILES:
ClC(=O)C1=CC=C(OCC2=CC=CC=C2)C=C1
1
–
1
of
1
results
1
Sigma Aldrich 4-Benzyloxybenzoylchloride
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.