accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14871-41-1

IUPAC Name: chloroiridiumylium; methanidylidyneoxidanium; bis(triphenylphosphane)
Synonyms: carbon monoxide; chloroiridiumylium; bis(triphenylphosphine)
Molecular Weight (g/mol): 780.26
Molecular Formula: C37H30ClIrOP2
InChi Key: PEPSDKMVHYXIMJ-UHFFFAOYSA-M
SMILES: Cl[Ir+].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)