Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbonylchlorobis(triphenylphosphine)iridium(I) 95.0+%, TCI America™
SDP

Catalog No. C2252200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2252200MG 200 mg
C22521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2252200MG Supplier TCI America Supplier No. C2252200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14871-41-1
Molecular Formula C37H30ClIrOP2
Molecular Weight (g/mol) 780.26
MDL Number MFCD00009590
InChI Key PEPSDKMVHYXIMJ-UHFFFAOYSA-M
Synonym carbonylchlorobis triphenylphosphine iridium i
PubChem CID 78076234
IUPAC Name chloroiridiumylium; methanidylidyneoxidanium; bis(triphenylphosphane)
SMILES Cl[Ir+].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 225°C
Color Yellow
Quantity 200 mg
UN Number 2811
Formula Weight 780.26
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Carbonylchlorobis(triphenylphosphine)iridium(I)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.