accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 148819-94-7

IUPAC Name: 2-(4-carboxylatophenyl)-1-hydroxy-4,4,5,5-tetramethyl-4,5-dihydro-1H-imidazol-3-ium-3-olate
Synonyms: 2-(4-carboxylatophenyl)-3-hydroxy-4,4,5,5-tetramethylimidazol-1-ium-1-olate
Molecular Weight (g/mol): 277.3
Molecular Formula: C14H17N2O4
InChi Key: QSSNLQPWTLQYTB-UHFFFAOYSA-M
SMILES: CC1(C)N(O)C(C2=CC=C(C=C2)C([O-])=O)=[N+]([O-])C1(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results