CAS RN 148839-33-2
IUPAC Name:
(5-chloro-2-methylphenyl)boronic acid
Synonyms:
5-chloro-2-methylphenylboronic acid
Molecular Weight (g/mol):
170.4
Molecular Formula:
C7H8BClO2
InChi Key:
QWTHTSAWMJFMOV-UHFFFAOYSA-N
SMILES:
CC1=CC=C(Cl)C=C1B(O)O