Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2-methylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. C26655G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2665-5G 5 g
C26651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2665-5G Supplier TCI America Supplier No. C26655G
Add to Cart
Add to Cart

Specifications

CAS 148839-33-2
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Molecular Formula C7H8BClO2
MDL Number MFCD03411939
Synonym 5-chloro-2-methylphenyl boronic acid, 5-chloro-2-methylphenylboronicacid, 5-chloro-2-methylbenzeneboronic acid, boronic acid, 5-chloro-2-methylphenyl, contains varying amounts of anhydride
InChI Key QWTHTSAWMJFMOV-UHFFFAOYSA-N
SMILES B(C1=C(C=CC(=C1)Cl)C)(O)O
IUPAC Name (5-chloro-2-methylphenyl)boronic acid
Molecular Weight (g/mol) 170.399
PubChem CID 2773336
Formula Weight 170.40
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.