accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COFFOH

CAS RN 148992-43-2

IUPAC Name: 1-ethoxy-2,2-difluoroethan-1-ol
Synonyms: 1-ethoxy-2,2-difluoroethanol
Molecular Weight (g/mol): 126.1
Molecular Formula: C4H8F2O2
InChi Key: WEEOMNFWRCDRJI-UHFFFAOYNA-N
SMILES: CCOC(O)C(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results