accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOOO

CAS RN 149-29-1

IUPAC Name: 4-hydroxy-2H,4H,6H-furo[3,2-c]pyran-2-one
Synonyms: patulin
Molecular Weight (g/mol): 154.12
Molecular Formula: C7H6O4
InChi Key: ZRWPUFFVAOMMNM-UHFFFAOYNA-N
SMILES: OC1OCC=C2OC(=O)C=C12
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)