Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Paromomycin sulfate, 98.3%, MP Biomedicals™

Catalog No. ICN15605605 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15605605 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15605605 Supplier MP Biomedicals, Inc Supplier No. 0215605605
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Broad-spectrum aminoglycoside antibiotic used to study bacterial protein synthesis and 30S ribosome assembly.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149-29-1 , 7732-18-5
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.12
InChI Key ZRWPUFFVAOMMNM-UHFFFAOYNA-N
IUPAC Name 4-hydroxy-2H,4H,6H-furo[3,2-c]pyran-2-one
SMILES OC1OCC=C2OC(=O)C=C12

Specifications

Type Paromomycin Sulfate
Physical Form Powder
Color Off-White
Quantity 5 g
Assay Percent Range >97%
Formula Weight 615.6
Percent Purity >97%
Grade Not Available
Chemical Name or Material Paromomycin Sulfate
Recommended Storage Store at room temperature (15° to 30°C)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.