accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOHOHOH

CAS RN 149-32-6

IUPAC Name: butane-1,2,3,4-tetrol
Synonyms: erythrite erythritol, meso- meso-eythritol i-erythritol C*eridex phycite lichen sugar phycitol butanetetrol (theta,S)-1,2,3,4-butanetetrol
Molecular Weight (g/mol): 122.12
Molecular Formula: C4H10O4
InChi Key: UNXHWFMMPAWVPI-UHFFFAOYNA-N
SMILES: OCC(O)C(O)CO
Boiling Point 
  • (2)
  • (5)
  • (3)
  • (3)
Quantity 
  • (1)
  • (4)
  • (4)
  • (1)
  • (3)
Physical Form 
  • (3)
  • (3)
  • (5)
Grade 
  • (1)
  • (2)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (1)
  • (5)
  • (3)
  • (5)
  • (2)
  • (5)
  • (3)
  • (3)
  • (1)
  • (4)
  • (4)
  • (1)
  • (3)
  • (3)
  • (3)
  • (5)
  • (1)
  • (2)
  • (5)
  • (3)
  • (5)