accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 149105-19-1

IUPAC Name: 2-(dihydroxyboranyl)benzoic acid
Synonyms: 2-(dihydroxyboranyl)benzoic acid
Molecular Weight (g/mol): 165.94
Molecular Formula: C7H7BO4
InChi Key: KWNPRVWFJOSGMZ-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC=C1C(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (4)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)