Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Carboxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. C25015G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2501-5G 5 g
C25011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2501-5G Supplier TCI America Supplier No. C25015G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 149105-19-1
Molecular Formula C7H7BO4
MDL Number MFCD01318118
InChI Key KWNPRVWFJOSGMZ-UHFFFAOYSA-N
PubChem CID 2733985

Specifications

Melting Point 162°C
Color White-Yellow
Quantity 5 g
Synonym 2-carboxyphenylboronic acid, 2-dihydroxyboranyl benzoic acid, 2-carboxybenzeneboronic acid, 2-dihydroxyboronyl benzoic acid, 2-dihydroxyboryl benzoic acid
SMILES OB(O)C1=CC=CC=C1C(O)=O
IUPAC Name 2-(dihydroxyboranyl)benzoic acid
Molecular Weight (g/mol) 165.94
Formula Weight 165.94
Physical Form Crystalline Powder
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.