accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNNHNNHOH2NNH(S)OOOOOCaO

CAS RN 1492-18-8

IUPAC Name: calcium (2S)-2-[(4-{[(2-amino-5-formyl-4-oxo-1,4,5,6,7,8-hexahydropteridin-6-yl)methyl]amino}phenyl)formamido]pentanedioate
Synonyms: Calcium folinate
Molecular Weight (g/mol): 511.51
Molecular Formula: C20H21CaN7O7
InChi Key: KVUAALJSMIVURS-AEQORCIRNA-L
SMILES: [Ca++].NC1=NC(=O)C2=C(NCC(CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC([O-])=O)C([O-])=O)N2C=O)N1
Molecular Weight (g/mol) 
  • (6)
  • (13)
Percent Purity 
  • (2)
  • (6)
  • (7)
  • (2)
Grade 
  • (1)
  • (13)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (7)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (13)
  • (6)
  • (1)
  • (2)
  • (7)
  • (2)
  • (2)
  • (7)
  • (6)
  • (13)
  • (2)
  • (6)
  • (7)
  • (2)
  • (1)
  • (13)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (7)
  • (2)