CAS RN 1492-30-4
IUPAC Name:
4-nitrophenyl hexadecanoate
Synonyms:
4-nitrophenyl hexadecanoate
Molecular Weight (g/mol):
377.53
Molecular Formula:
C22H35NO4
InChi Key:
LVZSQWIWCANHPF-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCC(=O)OC1=CC=C(C=C1)[N+]([O-])=O
1
–
2
of
2
results
1
Promotions Available
4-Nitrophenyl palmitate, 98+%
4-Nitrophenyl palmitate, 98+%
CAS: 1492-30-4 Molecular Formula: C22H35NO4 Molecular Weight (g/mol): 377.525 MDL Number: MFCD00047732 InChI Key: LVZSQWIWCANHPF-UHFFFAOYSA-N Synonym: 4-nitrophenyl palmitate,p-nitrophenyl palmitate,hexadecanoic acid 4-nitrophenyl ester,4-nitrophenyl hexadecanoate,hexadecanoic acid, 4-nitrophenyl ester,4-nitrophenylpalmitate,paranitrophenyl palmitate,para-nitrophenyl palmitate,p-nitrophenyl hexadecanoate,4-nitrophenyl palmitate # PubChem CID: 73891 ChEBI: CHEBI:85645 IUPAC Name: (4-nitrophenyl) hexadecanoate SMILES: CCCCCCCCCCCCCCCC(=O)OC1=CC=C(C=C1)[N+](=O)[O-]