Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Nitrophenyl Palmitate, MP Biomedicals™

Catalog No. ICN10088201 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
ICN10088201 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10088201 Supplier MP Biomedicals, Inc Supplier No. 0210088201

missing translation for 'validMainframeMsg'

Specifications

CAS 1492-30-4
Molecular Formula C22H35NO4
Quantity 1 g
Synonym 4-nitrophenyl palmitate, p-nitrophenyl palmitate, hexadecanoic acid 4-nitrophenyl ester, 4-nitrophenyl hexadecanoate, hexadecanoic acid, 4-nitrophenyl ester, 4-nitrophenylpalmitate, paranitrophenyl palmitate, para-nitrophenyl palmitate, p-nitrophenyl hexadecanoate, 4-nitrophenyl palmitate #
InChI Key LVZSQWIWCANHPF-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCCC(=O)OC1=CC=C(C=C1)[N+](=O)[O-]
IUPAC Name (4-nitrophenyl) hexadecanoate
Molecular Weight (g/mol) 377.525
PubChem CID 73891
ChEBI CHEBI:85645
Formula Weight 377.5
Physical Form Solid
Chemical Name or Material p-Nitrophenyl Palmitate
Show More Show Less
Recommended Storage Store at 0°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.