accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2H2N

CAS RN 14923-84-3

IUPAC Name: pyrene-1,6-diamine
Synonyms: pyrene-1,6-diamine
Molecular Weight (g/mol): 232.29
Molecular Formula: C16H12N2
InChi Key: OWJJRQSAIMYXQJ-UHFFFAOYSA-N
SMILES: NC1=CC=C2C=CC3=C(N)C=CC4=CC=C1C2=C34
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results