Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,6-Diaminopyrene 98.0+%, TCI America™
SDP

Catalog No. D2160100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2160100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D2160100MG Supplier TCI America Supplier No. D2160100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14923-84-3
Molecular Formula C16H12N2
Molecular Weight (g/mol) 232.29
MDL Number MFCD00142568
InChI Key OWJJRQSAIMYXQJ-UHFFFAOYSA-N
PubChem CID 146178
IUPAC Name pyrene-1,6-diamine
SMILES NC1=CC=C2C=CC3=C(N)C=CC4=CC=C1C2=C34

Specifications

Melting Point 230°C
Quantity 100 mg
Formula Weight 232.29
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,6-Diaminopyrene
RTECSNumber UR2451000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.