CAS RN 14936-66-4
IUPAC Name:
nonan-2-yl acetate
Synonyms:
nonan-2-yl acetate
Molecular Weight (g/mol):
186.3
Molecular Formula:
C11H22O2
InChi Key:
GSUGVJOUDSLEBL-UHFFFAOYNA-N
SMILES:
CCCCCCCC(C)OC(C)=O
1
–
1
of
1
results
1
2-Nonyl Acetate 98.0+%, TCI America™
CAS: 14936-66-4 Molecular Formula: C11H22O2 Molecular Weight (g/mol): 186.295 MDL Number: MFCD01321125 InChI Key: GSUGVJOUDSLEBL-UHFFFAOYSA-N Synonym: Acetic Acid 1-Methyloctyl Ester, 1-Methyloctyl Acetate, Acetic Acid 2-Nonyl Ester PubChem CID: 85788 IUPAC Name: nonan-2-yl acetate SMILES: CCCCCCCC(C)OC(=O)C