Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nonyl Acetate 98.0+%, TCI America™
SDP

Catalog No. A141925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A141925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A141925G Supplier TCI America Supplier No. A141925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14936-66-4
Molecular Formula C11H22O2
Molecular Weight (g/mol) 186.295
MDL Number MFCD01321125
InChI Key GSUGVJOUDSLEBL-UHFFFAOYSA-N
Synonym Acetic Acid 1-Methyloctyl Ester, 1-Methyloctyl Acetate, Acetic Acid 2-Nonyl Ester
PubChem CID 85788
IUPAC Name nonan-2-yl acetate
SMILES CCCCCCCC(C)OC(=O)C

Specifications

Color Yellow
Boiling Point 215°C
Quantity 25 g
Formula Weight 186.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Nonyl Acetate
EINECSNumber (2)-0735
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.