accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14937-45-2

IUPAC Name: tributyl(hexadecyl)phosphanium bromide
Synonyms: tributyl(hexadecyl)phosphanium bromide
Molecular Weight (g/mol): 507.67
Molecular Formula: C28H60BrP
InChi Key: RYVBINGWVJJDPU-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCCCCCCCCCC[P+](CCCC)(CCCC)CCCC
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)