Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tributylhexadecylphosphonium Bromide

Catalog No. 27620025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-620-025G 25 g
27-620-0100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 27-620-025G Supplier Sigma Aldrich Supplier No. 27620025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tributylhexadecylphosphonium Bromide

Specifications

CAS 14937-45-2
Melting Point 56°C to 58°C
Molecular Formula C28H60BrP
Linear Formula CH3(CH2)15P+[(CH2)3CH3]3Br-
MDL Number MFCD00011775
Quantity 25 g
Synonym Hexadecyltributylphosphonium bromide; TBHDPB
Molecular Weight (g/mol) 507.65 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.