accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNNH

CAS RN 149554-29-0

IUPAC Name: 6-(piperazin-1-yl)pyridine-3-carbonitrile
Synonyms: 6-(piperazin-1-yl)pyridine-3-carbonitrile
Molecular Weight (g/mol): 188.23
Molecular Formula: C10H12N4
InChi Key: ZYNPMKJQFWNFMI-UHFFFAOYSA-N
SMILES: N#CC1=CC=C(N=C1)N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results