accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14970-87-7

IUPAC Name: 2-[2-(2-sulfanylethoxy)ethoxy]ethane-1-thiol
Synonyms: 2-[2-(2-sulfanylethoxy)ethoxy]ethanethiol
Molecular Weight (g/mol): 182.3
Molecular Formula: C6H14O2S2
InChi Key: HCZMHWVFVZAHCR-UHFFFAOYSA-N
SMILES: SCCOCCOCCS
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)