accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14972-70-4

RhClClClClClClNaNaNaH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2O

CAS RN 14972-70-4

IUPAC Name: trisodium hexachlororhodiumheptakis(ylium) dodecahydrate
Synonyms: trisodium hexachlororhodiumheptakis(ylium) dodecahydrate
Molecular Weight (g/mol): 600.75
Molecular Formula: Cl6H24Na3O12Rh
InChi Key: JCMYDKHYUGZPJG-UHFFFAOYSA-H
SMILES: O.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].Cl[Rh+7](Cl)(Cl)(Cl)(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results