accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2OClClCr

CAS RN 14977-61-8

IUPAC Name: λ²-chromium(2+) dihydrate dichloride
Synonyms: chromic oxychloride
Molecular Weight (g/mol): 158.93
Molecular Formula: Cl2CrH4O2
InChi Key: HOCPFYCTFLRIAK-UHFFFAOYSA-L
SMILES: O.O.[Cl-].[Cl-].[Cr++]
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)